Assay Details
| Gene Symbol: RETSAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022404756.1 | XP_022260464.1 | 5 | 871 | 141 | |
| XM_540198.5 | XP_540198.1 | 5 | 1144 | 141 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RETSAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022404756.1 | XP_022260464.1 | 5 | 871 | 141 | |
| XM_540198.5 | XP_540198.1 | 5 | 1144 | 141 | ||
Set Membership: |
|