Assay Details
| Gene Symbol: CYLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005617567.3 | XP_005617624.1 | 15 - 16 | 2560 | 84 | |
| XM_005617568.3 | XP_005617625.1 | 14 - 15 | 2573 | 84 | ||
| XM_005617569.3 | XP_005617626.1 | 12 - 13 | 2357 | 84 | ||
| XM_849343.5 | XP_854436.1 | 15 - 16 | 2582 | 84 | ||
More Information
Set Membership: |
|

CYLD
Genomic Map