Assay Details
| Gene Symbol: SPACA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625181.2 | XP_005625238.1 | 4 | 651 | 138 | |
| XM_005625186.2 | XP_005625243.1 | 3 | 355 | 138 | ||
| XM_022423660.1 | XP_022279368.1 | 4 | 429 | 138 | ||
| XM_022423661.1 | XP_022279369.1 | 4 | 628 | 138 | ||
| XM_022423662.1 | XP_022279370.1 | 3 | 604 | 138 | ||
| XM_022423663.1 | XP_022279371.1 | 2 | 684 | 138 | ||
| XM_022423664.1 | XP_022279372.1 | 4 | 471 | 138 | ||
| XM_022423665.1 | XP_022279373.1 | 2 | 473 | 138 | ||
| XM_022423666.1 | XP_022279374.1 | 5 | 523 | 138 | ||
| XM_022423667.1 | XP_022279375.1 | 2 | 406 | 138 | ||
| XM_022423668.1 | XP_022279376.1 | 5 | 520 | 138 | ||
| XM_022423669.1 | XP_022279377.1 | 5 | 747 | 138 | ||
| XM_845436.5 | XP_850529.1 | 2 | 669 | 138 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SPACA9
Genomic Map