Assay Details
| Gene Symbol: SH3BP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022403798.1 | XP_022259506.1 | 1436 | 145 | ||
| XM_854895.5 | XP_859988.1 | 1271 | 145 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SH3BP5L| Gene Symbol: SH3BP5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022403798.1 | XP_022259506.1 | 1436 | 145 | ||
| XM_854895.5 | XP_859988.1 | 1271 | 145 | |||
Set Membership: |
|