Assay Details
| Gene Symbol: TGFB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_849026.5 | XP_854119.2 | 1 | 1151 | 59 | |
| GenBank mRNA | DQ310186.1 | - | 1 | 124 | 59 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TGFB3
Genomic Map