Assay Details
| Gene Symbol: NSUN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022414061.1 | XP_022269769.1 | 1034 | 179 | ||
| XM_843944.5 | XP_849037.1 | 1072 | 179 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NSUN3
Genomic Map