Assay Details
| Gene Symbol: GABRA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022426954.1 | XP_022282662.1 | 3 - 4 | 259 | 144 | |
| XM_022426955.1 | XP_022282663.1 | 3 - 4 | 259 | 144 | ||
| XM_022426956.1 | XP_022282664.1 | 2 - 3 | 201 | 144 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022426956.1 transcripts.
Set Membership: |
|

GABRA2
Genomic Map