Assay Details
| Gene Symbol: USP12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014107398.2 | XP_013962873.1 | 454 | 128 | ||
| XM_022409935.1 | XP_022265643.1 | 640 | 128 | |||
| XM_543159.6 | XP_543159.2 | 740 | 128 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

USP12
Genomic Map