Assay Details
| Gene Symbol: SPATA2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620541.2 | XP_005620598.1 | 434 | 67 | ||
| XM_845856.4 | XP_850949.2 | 629 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA2L| Gene Symbol: SPATA2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620541.2 | XP_005620598.1 | 434 | 67 | ||
| XM_845856.4 | XP_850949.2 | 629 | 67 | |||
Set Membership: |
|