Assay Details
| Gene Symbol: CCDC88B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631539.3 | XP_005631596.1 | 25 | 4091 | 90 | |
| XM_005631540.3 | XP_005631597.1 | 24 | 4260 | 90 | ||
| XM_014121339.2 | XP_013976814.1 | 26 | 4539 | 90 | ||
| XM_540883.6 | XP_540883.3 | 25 | 4238 | 90 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CCDC88B
Genomic Map