Assay Details
| Gene Symbol: ZNF23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022419234.1 | XP_022274942.1 | 4 | 869 | 145 | |
| XM_022419235.1 | XP_022274943.1 | 4 | 866 | 145 | ||
| XM_022419236.1 | XP_022274944.1 | 6 | 1378 | 145 | ||
| XM_022419237.1 | XP_022274945.1 | 7 | 1135 | 145 | ||
| XM_022419238.1 | XP_022274946.1 | 6 | 1428 | 145 | ||
More Information
Set Membership: |
|

ZNF23
Genomic Map