Assay Details
Gene Symbol: SHTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005637772.3 | XP_005637829.1 | 4 - 5 | 231 | 98 | |
XM_014108985.2 | XP_013964460.1 | 3 - 4 | 209 | 98 | ||
XM_014108986.2 | XP_013964461.1 | 3 - 4 | 209 | 98 | ||
XM_022411716.1 | XP_022267424.1 | 3 - 4 | 209 | 98 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
