Assay Details
| Gene Symbol: TUT7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005615793.2 | XP_005615850.1 | 6 - 7 | 1270 | 148 | |
| XM_005615794.3 | XP_005615851.1 | 6 - 7 | 1257 | 148 | ||
| XM_005615795.2 | XP_005615852.1 | 6 - 7 | 1270 | 148 | ||
| XM_005615796.2 | XP_005615853.1 | 6 - 7 | 1270 | 148 | ||
| XM_022420449.1 | XP_022276157.1 | 6 - 7 | 1137 | 148 | ||
| XM_533505.5 | XP_533505.2 | 6 - 7 | 1270 | 148 | ||
More Information
Set Membership: |
|

TUT7
Genomic Map