Assay Details
| Gene Symbol: RBM14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631423.3 | XP_005631480.1 | 3 | 1139 | 89 | |
| XM_847210.5 | XP_852303.1 | 3 | 1145 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: RBM14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631423.3 | XP_005631480.1 | 3 | 1139 | 89 | |
| XM_847210.5 | XP_852303.1 | 3 | 1145 | 89 | ||
Set Membership: |
|