Assay Details
| Gene Symbol: GALNT16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022422164.1 | XP_022277872.1 | 1 - 2 | 410 | 155 | |
| XM_022422165.1 | XP_022277873.1 | 1 - 2 | 411 | 155 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GALNT16
Genomic Map