Assay Details
| Gene Symbol: TTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005630443.3 | XP_005630500.1 | 5 | 808 | 106 | |
| XM_005630444.3 | XP_005630501.1 | 4 | 462 | 106 | ||
| XM_022404729.1 | XP_022260437.1 | 5 | 636 | 106 | ||
| XM_540180.6 | XP_540180.2 | 5 | 799 | 106 | ||
More Information
Set Membership: |
|

TTL
Genomic Map