Assay Details
| Gene Symbol: CDK15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014110900.2 | XP_013966375.1 | 5 - 6 | 1082 | 126 | |
| XM_022415194.1 | XP_022270902.1 | 5 - 6 | 1077 | 126 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CDK15
Genomic Map