Assay Details
| Gene Symbol: PACSIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627229.3 | XP_005627286.1 | 7 - 8 | 1085 | 143 | |
| XM_005627231.2 | XP_005627288.1 | 7 - 8 | 1196 | 143 | ||
| XM_005627233.3 | XP_005627290.1 | 7 - 8 | 1109 | 143 | ||
| XM_014118377.2 | XP_013973852.1 | 7 - 8 | 1102 | 143 | ||
| XM_014118378.2 | XP_013973853.1 | 7 - 8 | 1124 | 143 | ||
| XM_022425948.1 | XP_022281656.1 | 7 - 8 | 1063 | 143 | ||
| XM_844990.4 | XP_850083.1 | 7 - 8 | 1174 | 143 | ||
More Information
Set Membership: |
|

PACSIN1
Genomic Map