Assay Details
| Gene Symbol: SYNCRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003431650.3 | XP_003431698.1 | 8 - 9 | 1148 | 151 | |
| XM_005627594.2 | XP_005627651.1 | 8 - 9 | 1148 | 151 | ||
| XM_005627596.3 | XP_005627653.1 | 7 - 8 | 993 | 151 | ||
| XM_022426321.1 | XP_022282029.1 | 8 - 9 | 1148 | 151 | ||
| XM_022426322.1 | XP_022282030.1 | 8 - 9 | 1148 | 151 | ||
| XM_532223.6 | XP_532223.3 | 8 - 9 | 1211 | 151 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SYNCRIP
Genomic Map