Assay Details
Gene Symbol: LRRC4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631098.3 | XP_005631155.1 | 8 | 2807 | 127 | |
XM_005631099.3 | XP_005631156.1 | 8 | 2757 | 127 | ||
XM_005631100.3 | XP_005631157.1 | 7 | 2615 | 127 | ||
XM_005631102.3 | XP_005631159.1 | 3 | 3009 | 127 | ||
XM_005631103.3 | XP_005631160.1 | 2 | 2942 | 127 | ||
XM_014121147.2 | XP_013976622.1 | 9 | 2949 | 127 | ||
XM_022405301.1 | XP_022261009.1 | 9 | 3017 | 127 | ||
XM_022405302.1 | XP_022261010.1 | 8 | 2882 | 127 | ||
XM_022405304.1 | XP_022261012.1 | 2 | 1991 | 127 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
