Assay Details
| Gene Symbol: TRAPPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632939.3 | XP_005632996.1 | 3 | 480 | 122 | |
| XM_005632940.3 | XP_005632997.1 | 3 | 476 | 122 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TRAPPC5
Genomic Map