Assay Details
| Gene Symbol: HTR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637529.3 | XP_005637586.1 | 1 | 646 | 75 | |
| XM_534958.6 | XP_534958.2 | 1 | 643 | 75 | ||
| GenBank mRNA | AJ628248.1 | - | 1 | 408 | 75 | |
| AJ628249.1 | - | 1 | 408 | 75 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HTR7
Genomic Map