Assay Details
| Gene Symbol: ANKRD46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022426585.1 | XP_022282293.1 | 471 | 130 | ||
| XM_532285.5 | XP_532285.2 | 376 | 130 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ANKRD46
Genomic Map