Assay Details
| Gene Symbol: YTHDF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005638012.3 | XP_005638069.1 | 5 | 1237 | 113 | |
| XM_005638013.3 | XP_005638070.1 | 4 | 1344 | 113 | ||
| XM_005638014.3 | XP_005638071.1 | 4 | 1045 | 113 | ||
| XM_005638015.3 | XP_005638072.1 | 3 | 1027 | 113 | ||
| XM_022412228.1 | XP_022267936.1 | 5 | 1162 | 113 | ||
| XM_022412229.1 | XP_022267937.1 | 4 | 1249 | 113 | ||
| XM_852818.5 | XP_857911.1 | 4 | 1282 | 113 | ||
More Information
Set Membership: |
|

YTHDF3
Genomic Map