Assay Details
| Gene Symbol: TRIM16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022418606.1 | XP_022274314.1 | 3 - 4 | 685 | 138 | |
| XM_546637.5 | XP_546637.2 | 3 - 4 | 906 | 138 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

TRIM16
Genomic Map