Assay Details
Gene Symbol: LOC100685226 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022427692.1 | XP_022283400.1 | 8 | 2390 | 166 | |
XM_022427695.1 | XP_022283403.1 | 8 | 2406 | 166 | ||
XM_022427696.1 | XP_022283404.1 | 5 | 1954 | 166 | ||
XM_022427697.1 | XP_022283405.1 | 6 | 1991 | 166 | ||
XM_022427698.1 | XP_022283406.1 | 5 | 2046 | 166 | ||
XM_022427700.1 | XP_022283408.1 | 6 | 2077 | 166 | ||
XM_022427702.1 | XP_022283410.1 | 5 | 2049 | 166 | ||
XM_022427703.1 | XP_022283411.1 | 5 | 2045 | 166 | ||
XM_022427704.1 | XP_022283412.1 | 5 | 2052 | 166 | ||
XM_022427705.1 | XP_022283413.1 | 5 | 1906 | 166 | ||
XM_022427706.1 | XP_022283414.1 | 5 | 1902 | 166 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
