Assay Details
| Gene Symbol: CBFA2T3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620565.3 | XP_005620622.1 | 948 | 104 | ||
| XM_005620568.3 | XP_005620625.1 | 751 | 104 | |||
| XM_005620569.3 | XP_005620626.1 | 642 | 104 | |||
| XM_022419084.1 | XP_022274792.1 | 870 | 104 | |||
| XM_022419085.1 | XP_022274793.1 | 2329 | 104 | |||
More Information
Set Membership: |
|

CBFA2T3
Genomic Map