Assay Details
| Gene Symbol: ITPRID2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640411.3 | XP_005640468.2 | 14 - 15 | 3037 | 131 | |
| XM_014110786.2 | XP_013966261.1 | 11 - 12 | 2846 | 131 | ||
| XM_014110788.2 | XP_013966263.1 | 14 - 15 | 3040 | 131 | ||
| XM_022414834.1 | XP_022270542.1 | 14 - 15 | 2953 | 131 | ||
More Information
Set Membership: |
|

ITPRID2
Genomic Map