Assay Details
| Gene Symbol: LMX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022415368.1 | XP_022271076.1 | 7 | 889 | 157 | |
| XM_846259.5 | XP_851352.2 | 8 | 1243 | 157 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LMX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022415368.1 | XP_022271076.1 | 7 | 889 | 157 | |
| XM_846259.5 | XP_851352.2 | 8 | 1243 | 157 | ||
Set Membership: |
|