Assay Details
| Gene Symbol: RAB26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621723.2 | XP_005621780.1 | 6 - 7 | 664 | 232 | |
| XM_022420245.1 | XP_022275953.1 | 6 - 7 | 664 | 232 | ||
| XM_022420247.1 | XP_022275955.1 | 6 - 7 | 664 | 154 | ||
| XM_022420249.1 | XP_022275957.1 | 6 - 7 | 859 | 232 | ||
| XM_848555.3 | XP_853648.1 | 6 - 7 | 664 | 154 | ||
More Information
Set Membership: |
|

RAB26
Genomic Map