Assay Details
| Gene Symbol: SNRPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022416835.1 | XP_022272543.1 | 7 - 8 | 562 | 154 | |
| XM_843830.5 | XP_848923.1 | 7 - 8 | 705 | 154 | ||
More Information
Set Membership: |
|

SNRPA1
Genomic Map