Assay Details
| Gene Symbol: CTLA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003106.1 | NP_001003106.1 | 3 - 4 | 625 | 130 | |
| GenBank mRNA | AF143204.1 | - | 3 - 4 | 592 | 130 | |
| AF154843.1 | - | 3 - 4 | 625 | 130 | ||
| AF215893.1 | - | 3 - 4 | 608 | 130 | ||
| LC191194.1 | - | 3 - 4 | 592 | 130 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CTLA4
Genomic Map