Assay Details
| Gene Symbol: PA2G4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022424045.1 | XP_022279753.1 | 850 | 147 | ||
| XM_531629.5 | XP_531629.2 | 850 | 147 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PA2G4
Genomic Map