Assay Details
| Gene Symbol: CRYGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080901.2 | NP_001074370.1 | 3 | 511 | 87 | |
| GenBank mRNA | EF191196.1 | - | 3 | 513 | 87 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CRYGS
Genomic Map