Assay Details
| Gene Symbol: CYB5RL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005620260.3 | XP_005620317.1 | 1 | 84 | 66 | |
| XM_014113769.2 | XP_013969244.1 | 2 | 388 | 66 | ||
| XM_848129.4 | XP_853222.1 | 2 | 387 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005620260.3 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CYB5RL
Genomic Map