Assay Details
| Gene Symbol: PHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022411310.1 | XP_022267018.1 | 3 - 4 | 352 | 80 | |
| XM_022411311.1 | XP_022267019.1 | 3 - 4 | 352 | 80 | ||
| XM_022411312.1 | XP_022267020.1 | 3 - 4 | 352 | 80 | ||
| XM_022411313.1 | XP_022267021.1 | 3 - 4 | 352 | 80 | ||
| XM_022411314.1 | XP_022267022.1 | 3 - 4 | 352 | 80 | ||
| XM_022411315.1 | XP_022267023.1 | 3 - 4 | 352 | 80 | ||
More Information
Set Membership: |
|

PHC1
Genomic Map