Assay Details
| Gene Symbol: MGAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005625864.3 | XP_005625921.1 | 2 | 2948 | 161 | |
| XM_005625865.3 | XP_005625922.1 | 2 | 1794 | 161 | ||
| XM_014117348.2 | XP_013972823.1 | 2 | 2244 | 161 | ||
| XM_022424402.1 | XP_022280110.1 | 2 | 4776 | 161 | ||
| XM_022424403.1 | XP_022280111.1 | 4 | 2979 | 161 | ||
| XM_022424404.1 | XP_022280112.1 | 4 | 2984 | 161 | ||
| XM_022424405.1 | XP_022280113.1 | 3 | 2867 | 161 | ||
| XM_022424406.1 | XP_022280114.1 | 2 | 2829 | 161 | ||
| XM_022424407.1 | XP_022280115.1 | 2 | 2277 | 161 | ||
More Information
Set Membership: |
|

MGAT3
Genomic Map