Assay Details
| Gene Symbol: ZSWIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005635123.3 | XP_005635180.1 | 2 | 440 | 126 | |
| XM_022409350.1 | XP_022265058.1 | 2 | 338 | 126 | ||
| XM_022409351.1 | XP_022265059.1 | 2 | 434 | 126 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ZSWIM1
Genomic Map