Assay Details
| Gene Symbol: TRIM63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022413016.1 | XP_022268724.1 | 4 - 5 | 1087 | 139 | |
| XM_022413030.1 | XP_022268738.1 | 4 - 5 | 620 | 139 | ||
| XM_022413032.1 | XP_022268740.1 | 4 - 5 | 628 | 139 | ||
| XM_022413033.1 | XP_022268741.1 | 4 - 5 | 554 | 139 | ||
| XM_544488.6 | XP_544488.2 | 4 - 5 | 599 | 139 | ||
More Information
Set Membership: |
|

TRIM63
Genomic Map