Assay Details
| Gene Symbol: STRADB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640539.3 | XP_005640596.1 | 5 - 6 | 541 | 126 | |
| XM_022415028.1 | XP_022270736.1 | 5 - 6 | 519 | 126 | ||
| XM_536030.4 | XP_536030.2 | 5 - 6 | 538 | 126 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

STRADB
Genomic Map