Assay Details
| Gene Symbol: ETS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619567.3 | XP_005619624.1 | 1 | 1443 | 123 | |
| XM_005619568.3 | XP_005619625.1 | 1 | 1462 | 123 | ||
| XM_022418096.1 | XP_022273804.1 | 2 | 1412 | 123 | ||
| XM_022418097.1 | XP_022273805.1 | 1 | 1359 | 123 | ||
| XM_022418098.1 | XP_022273806.1 | 1 | 1289 | 123 | ||
| XM_022418099.1 | XP_022273807.1 | 2 | 1202 | 123 | ||
| XM_022418100.1 | XP_022273808.1 | 1 | 1542 | 123 | ||
| XM_022418101.1 | XP_022273809.1 | 2 | 1179 | 123 | ||
| XM_546405.5 | XP_546405.2 | 1 | 1440 | 123 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ETS1
Genomic Map