Assay Details
| Gene Symbol: KBTBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619843.3 | XP_005619900.1 | 3 | 1253 | 154 | |
| XM_022418365.1 | XP_022274073.1 | 4 | 1654 | 154 | ||
More Information
Set Membership: |
|

KBTBD3
Genomic Map