Assay Details
| Gene Symbol: RAB15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022422118.1 | XP_022277826.1 | 4 | 292 | 99 | |
| XM_022422119.1 | XP_022277827.1 | 4 | 927 | 99 | ||
More Information
Set Membership: |
|

RAB15
Genomic Map