Assay Details
| Gene Symbol: HMGB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001002937.2 | NP_001002937.1 | 320 | 125 | ||
| XM_022409535.1 | XP_022265243.1 | 327 | 125 | |||
| GenBank mRNA | AY135519.1 | - | 242 | 125 | ||
| DN349572.1 | - | 2 | 320 | 125 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HMGB1
Genomic Map