Assay Details
| Gene Symbol: PRKACB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622016.2 | XP_005622073.1 | 9 - 10 | 772 | 106 | |
| XM_005622017.3 | XP_005622074.1 | 9 - 10 | 961 | 106 | ||
| XM_005622018.3 | XP_005622075.1 | 8 - 9 | 744 | 106 | ||
| XM_005622019.3 | XP_005622076.1 | 8 - 9 | 937 | 106 | ||
| XM_005622020.3 | XP_005622077.1 | 7 - 8 | 912 | 106 | ||
| XM_022420584.1 | XP_022276292.1 | 8 - 9 | 937 | 106 | ||
| XM_537099.5 | XP_537099.1 | 7 - 8 | 986 | 106 | ||
| XM_862424.5 | XP_867517.1 | 7 - 8 | 736 | 106 | ||
| XM_862450.4 | XP_867543.1 | 7 - 8 | 784 | 106 | ||
More Information
Set Membership: |
|

PRKACB
Genomic Map