Assay Details
| Gene Symbol: DDAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621986.3 | XP_005622043.1 | 3 - 4 | 263 | 104 | |
| XM_022420551.1 | XP_022276259.1 | 3 - 4 | 278 | 104 | ||
| XM_547303.5 | XP_547303.1 | 3 - 4 | 479 | 104 | ||
More Information
Set Membership: |
|

DDAH1
Genomic Map