Assay Details
| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627579.3 | XP_005627636.1 | 27 - 28 | 6055 | 136 | |
| XM_005627580.3 | XP_005627637.1 | 26 - 27 | 6025 | 136 | ||
| XM_022426290.1 | XP_022281998.1 | 29 - 30 | 6411 | 136 | ||
| XM_022426291.1 | XP_022281999.1 | 28 - 29 | 6258 | 136 | ||
| XM_022426292.1 | XP_022282000.1 | 26 - 27 | 5900 | 136 | ||
| XM_022426293.1 | XP_022282001.1 | 26 - 27 | 5873 | 136 | ||
| XM_022426294.1 | XP_022282002.1 | 22 - 23 | 5252 | 136 | ||
| XM_022426295.1 | XP_022282003.1 | 22 - 23 | 5225 | 136 | ||
| XM_022426296.1 | XP_022282004.1 | 27 - 28 | 6059 | 136 | ||
| XM_022426297.1 | XP_022282005.1 | 17 - 18 | 4679 | 136 | ||
| XM_862572.5 | XP_867665.2 | 27 - 28 | 6028 | 136 | ||
More Information
Set Membership: |
|

DOP1A
Genomic Map