Assay Details
| Gene Symbol: TMEM79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421266.1 | XP_022276974.1 | 4 | 2044 | 177 | |
| XM_022421267.1 | XP_022276975.1 | 5 | 1495 | 177 | ||
| XM_022421268.1 | XP_022276976.1 | 5 | 1491 | 177 | ||
| XM_849873.3 | XP_854966.1 | 4 | 2040 | 177 | ||
More Information
Set Membership: |
|

TMEM79
Genomic Map