Assay Details
| Gene Symbol: CSNK2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005627149.3 | XP_005627206.1 | 4 - 5 | 453 | 129 | |
| XM_014118320.2 | XP_013973795.1 | 4 - 5 | 443 | 129 | ||
| XM_532075.6 | XP_532075.3 | 4 - 5 | 451 | 129 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CSNK2B
Genomic Map