Assay Details
| Gene Symbol: C7H1orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421373.1 | XP_022277081.1 | 4 - 5 | 750 | 143 | |
| XM_022421375.1 | XP_022277083.1 | 5 - 6 | 836 | 143 | ||
| XM_022421376.1 | XP_022277084.1 | 4 - 5 | 731 | 143 | ||
| XM_847193.4 | XP_852286.2 | 5 - 6 | 844 | 143 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

C7H1orf43
Genomic Map